Learn More
DWK Life Sciences Screw Cap
Screw caps provide a tight seal without the need for crimping and decapping tools.
£13.40 - £41.15
Specifications
For Use With (Application) High Performance Liquid Chromatography (HPLC)
Hardness 45 ± 5 shore A
Autoclavable Autoclavable
Quantity 100
View More Specs
Products 8
Product Code Brand Closure Size Type Color Material Price Quantity & Availability  
Product Code Brand Closure Size Type Color Material Price Quantity & Availability  
16676882
View Documents Promotion Details
DWK Life Sciences
225784
ND8 Open Top Cap Black White Silicone, Red PTFE, Polypropylene
£23.15
Pack of 100
Estimated Shipment: 28-02-2025
to see stock.
Add to basket
16686882
View Documents
DWK Life Sciences
225785
ND8 Open Top Cap Black White Silicone, Red PTFE, Pre-slit (single), Polypropylene
£41.15
Pack of 100
Estimated Shipment: 28-02-2025
to see stock.
Add to basket
16696882
View Documents
DWK Life Sciences
225786
ND8 Open Top Cap Black Red PTFE, White Silicone, Red PTFE, Polypropylene
£30.70
Pack of 100
Estimated Shipment: 28-02-2025
to see stock.
Add to basket
16626892
View Documents
DWK Life Sciences
225789
ND9 Open Top Cap Blue Red PTFE, White Silicone, Red PTFE, Polypropylene This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16646892
View Documents
DWK Life Sciences
225791
ND9 Open Top Cap White Red Rubber, Clear PTFE, Polypropylene
£13.40
Pack of 100
Estimated Shipment: 28-02-2025
to see stock.
Add to basket
16656892
View Documents
DWK Life Sciences
225792
ND9 Open Top Cap White White Silicone, Red PTFE, Polypropylene
£20.85
Pack of 100
Estimated Shipment: 28-02-2025
to see stock.
Add to basket
16666892
View Documents
DWK Life Sciences
225793
ND9 Open Top Cap White Red PTFE, White Silicone, Red PTFE, Polypropylene
£28.30
Pack of 100
Estimated Shipment: 28-02-2025
to see stock.
Add to basket
16606902
View Documents
DWK Life Sciences
225797
ND13 Open Top Cap Black White Silicone, Red PTFE, Polypropylene
£33.25
Pack of 100
Estimated Shipment: 21-01-2025
to see stock.
Add to basket
Description
Description
  • Packed in zip-lock PE-bags with tear-off foil strip to ensure tamper-evidence and re-closeability
Specifications
Specifications
High Performance Liquid Chromatography (HPLC)
45 ± 5 shore A
Autoclavable
100