missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Screw Cap Micro Tubes
GREENER_CHOICE

Product Code. 11529944 Shop All MBP Molecular BioProducts Products
Click to view available options
Capacity (Metric):
0.5 mL
1.5 mL
2 mL
Bottom Shape:
Conical
Non-skirted
Self-standing
Skirted
Closure Type:
Attached Cap
Not Included
Screw Cap with O-ring (Attached)
Screw Cap with O-ring (Unattached)
Packaging:
50 tubes and attached caps/bag, 10 bags/unit
50 tubes/bag, 10 bags/unit
50 tubes/bag, 10 bags/unit, 10 units/case; 500 caps/bag/unit
500 x pk, 10 pk x cs, 5000 x cs. Single Bagged. ZipLock bag.
500/Pack
Unit Size:
Pack of 500
Pack of 5000

Product Code. 11529944

Brand: Thermo Scientific™ 3474

Only null left
Add to basket
Add to basket

Choose these tubes, which available in conical and free-standing designs when seeking secure, short-term sample storage in low temperatures.

Choose Thermo Scientific™ Screw Cap Micro Tubes, which available in conical and free-standing designs, when seeking secure, short-term sample storage in low temperatures. Their robust design and material selection are well suited for work with biologics and offer rich features, including ultra-clear, externally threaded tubes and UV protective amber tubes. Options include: with or without a self-standing skirted bottom, with or without tethered straps, non-knurl and knurled tubes for single-hand operation, choice of multicolored caps and volume sizes of 0.5mL, 1.5mL and 2.0mL sizes. Screw Cap Micro Tube Caps offer a durable design incorporating an O-ring that helps prevent leaks under demanding conditions.

Non-Sterile is free of Rnase/Dnase, DNA, Pyrogen & Endotoxin.
Sterile E-Beam is free of Rnase/Dnase, DNA, Pyrogen, Endotoxin, ATP and PCR Inhibitor.
Packaging is tailored to meet broad requirements. Tubes are packaged with and without caps, caps are attached or unattached, placed in a single bag and double bagged feature for clean room environments is available upon request.
100% virgin polypropylene resin specifically selected for high clarity and biologic applications.
O-ring design, thickness and material selection have been optimized to provide protective features for maximum sealing ability, biologic interaction and temperature capability. The black colored O-ring is made from EPDM.
Automated System Compatible. Whether automate is used for filling, capping or labeling, the product is designed to withstand high torque pressures associated with automation. Product design helps to address common automated issues like: fissure cracks, bowl feeder jam, over/under torqued caps and cross threading.
Volume range of 0.5, 1.5 and 2.0mL.
Tube Color choices are an ultra-high clarity, natural color or amber option which provides ultra violet (UV) protection of 1.22% transmittance between 220 and 500nm.
Standard or Tethered cap, all available in a rainbow of colors. Visit cap product page for additional information.
Knurl tubes feature the ability to lock tube into specialty racks for single-hand operation.
Non knurl tubes ideal for automation and labeling, tubes are plain, removed of any labeling interference.
Skirted or Self Standing tubes can stand upright on their own, no rack required.
Conical tubes requires a rack to support them in the upright position.
Low nucleic acid and protein binding and low enzyme inhibition are features that Resin selection features.
RCF range from 18,000 to 25,000 x g.
Temperature Range -80°C to +95°C. Short-term storage in -80°C, -20°C, dry ice and wet ice. Short-term storage in vapor phase LN2. Suitable for a quick dip flash freeze in LN2.
Water Bath compatible up to +95°C.

TRUSTED_SUSTAINABILITY

Specifications

Autoclavable Not Autoclavable
Capacity (Metric) 1.5 mL
Color Natural
Material Polypropylene (Virgin)
Includes Knurl
Packaging 500 x pk, 10 pk x cs, 5000 x cs. Single Bagged. ZipLock bag.
Temperature Range (Metric) Cold Storage: -80° to +95°C. Liquid Nitrogen: Short term Vapor Phase & Quick Flash Freeze acceptable, not recommmended for LN2 submersion storage
Clean Claims Non-Sterile Free of: Rnase/Dnase, Human and Mouse DNA, Pyrogen & Endotoxin, ATP, PCR Inhibitor
Binding Property Low Nucleic Acid and Protein Binding & Low Enzyme Inhibition
Max. Torque Manual/Hand Torque: Standard: 0.125 lbsf.in to 2.5 lbsf.in; Metric: 1.412 Ncm to 28.24 Ncm. Automated/Robotic Torque Standard: 1.312 lbsf.in to 2.5 lbsf.in; Metric: 14.83 Ncm to 28.24 Ncm
Sterility Non-sterile
Graduated Yes
Closure Type Screw Cap with O-ring (Unattached)
Fit for Purpose PCR Ready
Bottom Shape Skirted
Max. RCF 18,000 x g
Product Type Screw-cap Microtube
Show More Show Less

† Subject to Thermo Fisher Scientific standard limited warranty. See thermoscientific.com or your sales representative for details.

Screw cap micro tubes and caps are not recommended for autoclave. Sterile options available.

Not intended to submerge and store in liquid phase LN2.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.