missing translation for 'onlineSavingsMsg'
Learn More

Labcon™ Screw Caps with Elastomeric Seal for SuperClear™ Microtubes

These screw caps with elastomeric seals are available in a wide range of colors.

Brand:  Labcon™ 3602-877-000-9

 View more versions of this product

Product Code. 17527916

  • £77.65 / Pack of 500
Estimated Shipment: 02-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • For SuperClear™ microcentrifuge tubes
  • With integral elastomeric seal
  • Cap includes soft gripping surface
  • Endotoxin-free (non-pyrogenic)
  • Polyethylene USP Class VI resin
  • Made in USA
Specifications

Specifications

12.7 mm
Blue
6.1 mm
5000/Cs.
Non-sterile
Polypropylene
Bagged
500/Pk.
500 Caps/Pk., 10 Pk./Cs.
0.5 to 2 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Special Offers

Special Offers