Learn More
Lenz Laborglasintrumente Screw Caps for Screwthread Tubes
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand: Lenz Laborglasintrumente 01320032
Additional Details : Weight : 0.10000kg
Description
- With DIN thread (GL)
- Temperature resistant up to 180 °C
- With PTFE coated silicone sealing (VMQ)
- Material: PBT
- Color: red
Specifications
PBT | |
Red | |
GL 32 |