missing translation for 'onlineSavingsMsg'
Learn More

Kautex™ Screw Closures

Product Code. 10095851
Click to view available options
Diameter (Metric):
28 mm
45 mm
80 mm
For Use With (Equipment):
1500/2500/4000 mL Bottle
250/350 mL Bottle
50 mL Bottle

Manufactured with polypropylene for excellent chemical resistance. Kautex™ Screw Closures are rugged caps designed for Kautex™ wide mouth HDPE bottles.

  • UN approval for solids when closures are used with associated bottles
  • Tamper evident ring
  • Conical seal provides leakproof closure
  • Available to fit bottles with capacities from 50 to 4000 mL
TRUSTED_SUSTAINABILITY

Specifications

Color Blue
Includes Tamper Evident Ring and Conical Seal
Product Type Screw Cap
For Use With (Equipment) 1500/2500/4000 mL Bottle
Diameter (Metric) 80 mm
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'