Learn More
Thermo Scientific™ Screws for Benchtop Shakers and Mixers
Get assorted accessory screws for Benchtop Shakers and mixers
Brand:  Thermo Scientific™ 88882130
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 16554812
  • £4.99 / Each
In Stock
to see stock.
Add to basket
Description
Description
Assorted accessory screws for benchtop shakers and mixers
Service Plan
Standard Thermo warranty guideline, provide repair service and free spare part within warranty
Services
2 years
Specifications
Specifications
Screw GB/T819.1 M4x8
Screw GB/T819.1 M4x8 for Digital Microplate shaker, Digital mini Rotator
4 screws
Stainless Steel
Digital microplate shaker, digital mini rotator
M4 x 8