missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Screw-Top Bottle, Square, Clear, Soda-Lime Glass, Narrow Mouth, High form DIN thread

Product Code. 11776918
Click to view available options
Capacity (Metric):
100 mL
250 mL
500 mL
1000 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

Square clear soda lime glass bottles, narrow mouth.

Square clear soda lime (Type 3) glass bottles, narrow mouth, high form DIN thread.

  • Square shape for efficient packing
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 250 mL
For Use With (Application) Storage of samples and reagents; Sample collection; Transport and shipping
Material Soda Lime Glass
Quantity 10/Pk.
Thread Size (Metric) GL 32
Autoclavable Not Autoclavable
Diameter (Metric) 64 mm
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'