Learn More
Thermo Scientific™ Screw Vial Convenience Kit
Store your samples in these vials to eliminate leaching of ions while maintaining a consistent pH for the life of your sample. Storage vials range from 2mL to 40mL.
£46.89 - £159.23
Product Code | Brand | Includes | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Includes | Price | Quantity & Availability | |||||
16595874
|
Thermo Scientific™
B7800-1-9A |
2mL amber glass vial, solid top PP screw cap |
£56.65
Pack of 100 |
Estimated Shipment: 03-10-2024
Log in to see stock. |
|||||
12428666
|
Thermo Scientific™
B7990-2 |
4mL clear glass vial, open-top cap |
£46.89
Pack of 100 |
Estimated Shipment: 12-09-2024
Log in to see stock. |
|||||
16585874
|
Thermo Scientific™
B7800-1-9 |
2mL clear glass vial, solid top PTFE-lined cap |
£72.29
Pack of 100 |
Estimated Shipment: 03-10-2024
Log in to see stock. |
|||||
12408666
|
Thermo Scientific™
B7990-12A |
12mL amber glass, open top PTFE/silicone septum |
£159.23
Pack of 200 |
Estimated Shipment: 09-09-2024
Log in to see stock. |
|||||
12468666
|
Thermo Scientific™
B7990-4 |
16mL clear glass vial, open top PP cap, PTFE/silicone septum |
£144.57
Pack of 200 |
In Stock
Log in to see stock. |
|||||
12448666
|
Thermo Scientific™
B7990-3 |
8mL clear glass vial with open top septa caps |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
12498656
|
Thermo Scientific™
B7990-12 |
12mL clear glass vial, open top PP cap, PTFE/silicone septum |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Store your samples in these vials to eliminate leaching of ions while maintaining a consistent pH for the life of your sample. Thermo Scientific™ National™ and Thermo Scientific™ Chromacol™ Sample Storage Screw Thread Vials are manufactured from superior quality 33 expansion borosilicate clear (Type 1, Class A) or 51A amber (Type 1, Class B) glass. Storage vials range from 2mL to 40mL.
- PTFE-lined solid-top storage caps
- PTFE on film/foam backing offers broad chemical resistance
- Not autoclavable
Unassembled convenience kits saves time during sample preparation.
- Solid-top convenience kits include matched quantities of vials and screw caps with pre-assembled septa.
- Open-top convenience kits contain shrink-wrapped vials and separately packaged caps and septa in polybags.
Specifications
Glass |