Learn More
Lenz Laborglasinstrumente™ Screwthread Tubes for Glassblowers
Manufactured from borosilicate glass. Lenz Laborglasinstrumente™ Screwthread Tubes for Glassblowers has a high resistance to thermal shock and chemical corrosion. The tube combined with plastic fittings produce a tight connection to a straight tube.
Brand: Lenz Laborglasinstrumente™ 01300032
Description
- Acc. To DIN 12216 and in addition to standar
- With DIN thread (GL)
- Material: DURAN™
Specifications
DIN 12216 | |
Borosilicate Glass (3.3) | |
2.0 mm | |
32 |
28 mm | |
140 mm | |
10 |