missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SD 1008, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
JAK2/STAT3 signaling pathway inhibitor. Inhibits activation of STAT3, JAK2 and Src. Induces apoptosis in cell lines expressing constitutively active tyrosine-phosphorylated STAT3.
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 329.35 |
Percent Purity | >99% |
Chemical Name or Material | SD 1008 |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'