missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SDHB Mouse anti-Human, Unconjugated, Clone: 1A2A5, Proteintech
Mouse Monoclonal Antibody
£135.00 - £349.00
Specifications
Antigen | SDHB |
---|---|
Clone | 1A2A5 |
Concentration | 1000 μg/mL |
Applications | Western Blot, Immunohistochemistry, Immunofluorescence, ELISA |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16880316
|
Proteintech
67600-1-IG-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16870316
|
Proteintech
67600-1-IG-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The SDHB antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human SDHB. This antibody recognizes human, mouse, rat antigen. The SDHB antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
SDHB | |
1000 μg/mL | |
Monoclonal | |
Mouse | |
6390 | |
Primary |
1A2A5 | |
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
Unconjugated | |
Human, Rat, Mouse | |
IgG1 |