Learn More
Cytiva SDS-PAGE Clean-up Kit
Prepares samples for SDS-PAGE which are difficult to analyze due to salts or low protein concentrations
Brand: Cytiva 80-6484-70
Code : GE
Additional Details : Weight : 0.69000kg Transport : UN number : 3316 Chem class : 9 Pack group : II
Includes: Precipitant,15mL; Co-precipitant, 15mL; Wash buffer, 50mL; Wash additive, 250μL; Buffer I, 2mL; Buffer II, 500μL; Sample buffer, 2.5mL
Description
- Quantitatively precipitates proteins while leaving interfering substances in solution
- Delivers quantitative yield in less than two hours
Specifications
219° to 220°C | |
SDS PAGE Clean-up Kit | |
15mL Precipitant, 15mL coprecipitant, 50mL wash Buffer, 250μL wash additive, 2mL Buffer I, 500μL Buffer II, 2.5mL sample Buffer, quick reference card, instructions | |
121.13 | |
Room Temperature |
Colorless | |
Liquid | |
168.5°C | |
1 Kit | |
50 samples of less than 100μL |
Safety and Handling
- Danger
Hazard Category
- Flammable liquid Category 3
- Serious eye damage/eye irritation Category 2
- Carcinogenicity Category 2
- Reproductive toxicity Category 2
- Specific target organ toxicity after repeated exposure Category 2
- LONG-TERM AQUATIC HAZARD Chronic 1
- Skin corrosion/irritation Category 1 B
- Skin corrosion/irritation Category 1 A
- Serious eye damage/eye irritation Category 1
- Specific target organ toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
- H226-Flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H351-Suspected of causing cancer.
- H361d-Suspected of damaging the unborn child.
- H336-May cause drowsiness or dizziness.
- H373-May cause damage to organs.
- H410-Very toxic to aquatic life with long lasting effects.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H335-May cause respiratory irritation.
- H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
- P201-Obtain special instructions before use.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P273-Avoid release to the environment.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P310-Immediately call a POISON CENTER/doctor/.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P331-Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305-IF IN EYES:.
- P235-Keep cool.
- P405-Store locked up.
- P501-Dispose of contents/container to .
Supplemental information
- MIXTURE LIST-Contains : sodium hydroxide, trometamol, sodium carbonate, Cholan-24-oic acid, 3, 12-dihydroxy-, monosodium sal