missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SDS Solution, 20% Sodium Dodecyl Sulfate Solution, Molecular Biology/Electrophoresis, Fisher BioReagents™
Click to view available options
Quantity:
200 mL
1 L
Packaging:
Nalgene Poly Bottle
Poly Bottle
Description
SDS Solution, 20% Sodium Dodecyl Sulfate Solution, Molecular Biology/Electrophoresis
- DNase, RNase, and protease-free premixed solution containing 20% SDS for protein extraction and DNA extraction procedures
- Reduce preparation time by using this premade SDS solution
- SDS is a commonly used detergent in protein purification, polyacrylamide gel electrophoresis (PAGE), and other molecular biology applications
- SDS is also used to disrupt cell membranes and dissociate nucleic acid-protein complexes in DNA extraction procedures
- Intended for research use only; not for human or animal diagnostic purposes

Specifications
Specifications
Absorbance | 0.4 max. (0.1% solution) at 230nm, 0.1 max. (0.1% solution) at 280nm |
Color | Colorless |
Physical Form | Liquid |
Boiling Point | 100°C |
Density | 1.01g/cm³ |
Concentration | 19.5 to 20.5% |
Quantity | 200 mL |
Identification | Pass Test |
Packaging | Poly Bottle |
Assay Percent Range | (as Fatty Alcohol Sulfate): ≥99% |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Safety and Handling

Product Identifier
- Sodium dodecyl sulfate, 10% solution
- Warning
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 2
- H318-Causes serious eye damage.
- H315-Causes skin irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Special Offers
-
Multiple Manufacturers
Chromatography and Chemicals Promotion
Explore our new promotion and get up to 70% off some of our most popular products.Promo Code: 24127Expires: 2025-05-31
New Offer Save Up to 70%
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
SDS Solution, 20% Sodium Dodecyl Sulfate Solution, Molecular Biology/Electrophoresis, Fisher BioReagents™ >
Spot an opportunity for improvement?Share a Content Correction