Learn More
Applied Biosystems™ SDS Spectral Calibration Kit
This kit enables users to calibrate the ABI PRISM 7700™ Sequence Detection System quickly and easily.
Brand:  Applied Biosystems™ 4305822
388.30 GBP valid until 2024-12-31
Use promo code "21651" to get your promotional price.
Product Code. 10269764
  • £706.00 / Each
Explore more special offers
For Research Use Only. All usage must comply with product instructions.
Description
Description
This easy-to-use kit establishes the pure dye spectra and multicomponent values for FAM™, JOE™, NED™, ROX™, SYBR™ Green, VIC™, TAMRA™, and TET™ dyes on the ABI PRISM 7700™ Sequence Detection System. The SDS Spectral Calibration kit includes 8 tubes of pure dye standards for the Applied Biosystems™ ABI PRISM 7700™ Sequence Detection System. Spend Your Time on Research - Not Instrument Calibration This kit enables users to calibrate the ABI PRISM 7700™ Sequence Detection System quickly and easily. The system automatically collects the dye standard's spectral information as part of the instrument installation procedure. The system's application algorithm then uses the calibration information to ensure accurate data analysis.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Includes the Passive Reference I dye standard for signal normalization in all real-time and endpoint reactions. Includes 8 tubes of pure dye standards (FAM™, JOE™, NED™,ROX™, SYBR™ Green, VIC™, TAMRA™, TET™), for ABI PRISM™ 7700 Sequence Detection System.
Spectral Calibration Kit
Tubes
Spectral Calibration
7700 System
Real time PCR (qPCR)
8 Tubes