missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Se-methylseleno-L-cysteine, 98%

Product Code. 10125370
Click to view available options
Quantity:
50 mg
250 mg
1 g
Packaging:
Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26046-90-2
Molecular Formula C4H9NO2Se
Molecular Weight (g/mol) 182.092
MDL Number MFCD00800565
InChI Key XDSSPSLGNGIIHP-VKHMYHEASA-N
Synonym methylselenocysteine, se-methylselenocysteine, se-methyl-l-selenocysteine, 3-methylseleno-l-alanine, selenium-methylselenocystine, r-2-amino-3-methylselanyl propanoic acid, se-methyl-seleno-l-cysteine, unii-twk220499z, ccris 5465, l-alanine, 3-methylseleno
PubChem CID 147004
ChEBI CHEBI:27812
IUPAC Name (2R)-2-amino-3-methylselanylpropanoic acid
SMILES C[Se]CC(C(=O)O)N

Specifications

Color White to Yellow
Assay Percent Range 98%
Infrared Spectrum Authentic
Packaging Glass Bottle
Formula Weight 182.08
Percent Purity ≥97.5%
Quantity 250 mg
Physical Form Powder
Chemical Name or Material Se-methylseleno-L-cysteine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'