Learn More
Thermo Scientific Chemicals Se-methylseleno-L-cysteine, 98%
£73.00 - £820.00
Chemical Identifiers
CAS | 26046-90-2 |
---|---|
Molecular Formula | C4H9NO2Se |
Molecular Weight (g/mol) | 182.092 |
MDL Number | MFCD00800565 |
InChI Key | XDSSPSLGNGIIHP-VKHMYHEASA-N |
Synonym | methylselenocysteine, se-methylselenocysteine, se-methyl-l-selenocysteine, 3-methylseleno-l-alanine, selenium-methylselenocystine, r-2-amino-3-methylselanyl propanoic acid, se-methyl-seleno-l-cysteine, unii-twk220499z, ccris 5465, l-alanine, 3-methylseleno |
PubChem CID | 147004 |
ChEBI | CHEBI:27812 |
IUPAC Name | (2R)-2-amino-3-methylselanylpropanoic acid |
SMILES | C[Se]CC(C(=O)O)N |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10096383
|
Thermo Scientific Chemicals
300640500 |
50 mg | Glass Bottle |
£73.00
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10125370
|
Thermo Scientific Chemicals
300642500 |
250 mg | Glass Bottle |
£248.00
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10167463
|
Thermo Scientific Chemicals
300640010 |
1 g | Glass Bottle |
£820.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
26046-90-2 | |
182.092 | |
XDSSPSLGNGIIHP-VKHMYHEASA-N | |
147004 | |
(2R)-2-amino-3-methylselanylpropanoic acid |
C4H9NO2Se | |
MFCD00800565 | |
methylselenocysteine, se-methylselenocysteine, se-methyl-l-selenocysteine, 3-methylseleno-l-alanine, selenium-methylselenocystine, r-2-amino-3-methylselanyl propanoic acid, se-methyl-seleno-l-cysteine, unii-twk220499z, ccris 5465, l-alanine, 3-methylseleno | |
CHEBI:27812 | |
C[Se]CC(C(=O)O)N |
Specifications
26046-90-2 | |
98% | |
Authentic | |
methylselenocysteine, se-methylselenocysteine, se-methyl-l-selenocysteine, 3-methylseleno-l-alanine, selenium-methylselenocystine, r-2-amino-3-methylselanyl propanoic acid, se-methyl-seleno-l-cysteine, unii-twk220499z, ccris 5465, l-alanine, 3-methylseleno | |
C[Se]CC(C(=O)O)N | |
182.092 | |
CHEBI:27812 | |
≥97.5% | |
Se-methylseleno-L-cysteine |
White to Yellow | |
C4H9NO2Se | |
MFCD00800565 | |
XDSSPSLGNGIIHP-VKHMYHEASA-N | |
(2R)-2-amino-3-methylselanylpropanoic acid | |
147004 | |
182.08 | |
Powder |
Safety and Handling
GHS H Statement:
Toxic if swallowed.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Very toxic to aquatic life with long lasting effects.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
RUO – Research Use Only