missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva SE260/SE250 MiniVertical Electrophoresis System Replacement Parts: Spacers
£60.12
Specifications
For Use With (Equipment) | SE 250, SE 260 Mini Format Vertical Gel Electrophoresis Unit |
---|---|
Type | T-shape Spacer |
Length (Metric) | 1.05 cm |
Product Code | Brand | Thickness (English) | Thickness (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Thickness (English) | Thickness (Metric) | Price | Quantity & Availability | ||||
10065794
|
Cytiva
SE2619T-2-1.5 |
0.05 in. | 0.15 cm |
£60.12
Pack of 2 |
In Stock
Log in to see stock. |
||||
10668285
|
Cytiva
SE2619T-2-1.0 |
0.03 in. | 0.1 cm |
£60.12
Pack of 2 |
In Stock
Log in to see stock. |
||||
Description
Specifications
SE 250, SE 260 Mini Format Vertical Gel Electrophoresis Unit | |
T-shape Spacer | |
1.05 cm |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title