Learn More
Cytiva SE260/SE250 MiniVertical Electrophoresis System Replacement Parts: Spineless Combs
Brand: Cytiva SE211A-15-1.0
74.31 GBP valid until 2024-12-31
Use promo code "21631" to get your promotional price.
This item is not returnable.
View return policy
Description
Specifications
SE 250, SE 260 Mini Format Vertical Gel Electrophoresis Unit | |
PTFE | |
0.03 in. | |
2.9μL/ mm | |
0.03 in. |
Spineless Comb | |
15 | |
0.1 cm | |
0.29 cm | |
0.1 cm |