missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hoefer™ SE640 Wide-Mini Vertical Electrophoresis Unit
Wide-mini system, ideal for rapid screening of multiple samples
£1081.34 - £1277.23
Specifications
Certifications/Compliance | EN61010-1, CSA C22.2 1010.1, CE |
---|---|
Humidity Range | Up to 80% |
Temperature (English) Maximum | 113°F |
Temperature (Metric) Maximum | 45°C |
Temperature (Metric) Storage | 15°C to 30°C |
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
11915358
|
Hoefer™
SE640-15-1.5 |
Wide-Mini Vertical Electrophoresis Unit, Complete |
£1277.23
Each |
Estimated Shipment: 07-11-2024
Log in to see stock. |
|||||
11985408
|
Hoefer™
SE640 |
Wide-Mini Vertical Electrophoresis Unit, Basic |
£1081.34
Each |
Estimated Shipment: 29-11-2024
Log in to see stock. |
|||||
Description
The Hoefer Wide-Mini Vertical Electrophoresis unit offers extra wide format ideal for quick, high throughput runs. With accessory divider plate you can complete a run of up to 112 samples in as little as 40 minutes.
- Maximum glass plate size 18 x 8 cm, permits rapid screening of up to 112 samples at one time
- Design permits gels to be surrounded uniformly by buffer so heat dissipation is efficient; bands are sharp and lanes are straight
- Accommodates denaturing or native polyacrylamide gels
- Run up to four second dimension gels suing IEF strips or tube gels
- Ideal for optimizing 2DE protocols
- Storage temperature: +15°C to 30°C
Specifications
EN61010-1, CSA C22.2 1010.1, CE | |
113°F | |
15°C to 30°C | |
7.08 in. | |
18 cm | |
3.14 in. | |
5.51 in. | |
1000 V | |
500 mA |
Up to 80% | |
45°C | |
12.59 in. | |
32 cm | |
11.41 in. | |
29 cm | |
14 cm | |
50 w |