missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sea Sand, Extra Pure, SLR, Fisher Chemical™
Click to view available options
Quantity:
1 kg
5 kg
Description
O2Si, CAS Number-14808-60-7, aerosil, cristobalite, diatomaceous earth, infusorial earth, quartz, silica, silica, amorphous, silicic anhydride, silicon dioxide, tridymite, 5kg, 2230 deg.C, CHEBI:30563, Beige, 60.08g/mol, VYPSYNLAJGMNEJ-UHFFFAOYSA-N, dioxosilane, 1610 deg.C, 11232, 60.083

Specifications
Specifications
Boiling Point | 2230°C |
Melting Point | 1610°C |
Color | Beige |
Physical Form | Solid |
Quantity | 1 kg |
Formula Weight | 60.08g/mol |
Packaging | Plastic powder jar |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction