Learn More
Lonza™ SeaKem™ LE Agarose
For analysis of nucleic acids by gel electrophoresis and blotting. Lonza SeaKem™ LE Agarose is recommended for protein gel electrophoresis in applications such as ouchterlony and radial immunodiffusion (RID).
Brand: Lonza™ 50005
Additional Details : Weight : 1.00000kg
Description
- Each gel sharply resolves DNA and provides consistent resolution from lot-to-lot
- Agarose has no detectable DNase or RNase activity
- Forms strong gels with low background upon ethidium bromide, SYBR™ Green, or GelStar™ staining
- Certificate of analysis is provided
Specifications
Agarose Gel Cassette | |
Ouchterlony and radial immunodiffusion (RID) | |
1 kg |