missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ Seal Crimper

Product Code. 15103075 Shop All Labconco Corporation Products
Click to view available options
Diameter (Metric) Outer:
13 mm
20 mm
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15103075

Brand: Labconco™ 7578100

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Secure tear-away aluminum seals.

  • Secure tear-away Aluminum Seals on Serum Bottles
  • Available in 13mm or 20mm corkage

Specifications

Diameter (Metric) Outer 20 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.