missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Labconco™ Seal Crimper
Secure tear-away aluminum seals.
Brand: Labconco™ 7578000
Code : 50
Additional Details : Weight : 1.36000kg
Description
- Secure tear-away Aluminum Seals on Serum Bottles
- Available in 13mm or 20mm corkage
Specifications
13 mm |