missing translation for 'onlineSavingsMsg'
Learn More

Mettler Toledo™ Sealing Discs

Product Code. 10611141 Shop All Mettler Toledo Products
Click to view available options
For Use With (Equipment):
Stromboli KF Oven Sample Changer Sample Vial
Unit Size:
Pack of 144

Product Code. 10611141

Brand: Mettler Toledo™ 51108693

This item requires a price update prior to being ordered.

Request a Price for this item here.

Suitable sealing discs for Stromboli KF Oven Sample Changer sample vials. Mettler Toledo™ Sealing Discs are made of aluminum and come in a pack of 144.

  • Suitable for the Stromboli KF Oven Sample Changer vials
  • Aluminum
  • 144 vials/pack
TRUSTED_SUSTAINABILITY

Specifications

Color Silver
Diameter (Metric) 22 mm
Type Sealing Discs
For Use With (Application) Stromboli KF Oven Sample Changer sample vial
Material Aluminum
For Use With (Equipment) Stromboli KF Oven Sample Changer Sample Vial
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.