Learn More
DWK Life Sciences Sealing Ring, for polished spherical joints (VITON)
For sealing ground joints without ground joints DURAN™, especially for vacuum applications.
Specifications
Material Viton
For Use With (Application) Sealing of non-sanded spherical ground joints
Quantity 10/Pk.
View More Specs
Products 4
Product Code Brand Joint Size Price Quantity & Availability  
Product Code Brand Joint Size Price Quantity & Availability  
15804498
View Documents
DWK Life Sciences
1155355
13 NS N/A
N/A
15814498
View Documents
DWK Life Sciences
1155356
19 NS N/A
N/A
15824498
View Documents
DWK Life Sciences
1155357
64 NS N/A
N/A
15894488
View Documents
DWK Life Sciences
1152556
29 NS N/A
N/A
Description
Description
Ball joints without ground joints only seal via the sealing ring. VITON sealing rings are used in vacuum applications.
  • Due to the material properties, it can be used for work with high demands on the chemical resistance of the sealing ring.
  • Sealing ring made of VITON
Specifications
Specifications
Viton
Sealing of non-sanded spherical ground joints
10/Pk.