missing translation for 'onlineSavingsMsg'
Learn More

Fisher Science Education™ Seamless Cellulose Dialysis Tubing

Retains materials with 12,000 and higher molecular weight.

Brand:  Fisher Science Education™ S25645A

 View more versions of this product

Product Code. 17780245

  • £16.20 / 1 roll

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Especially useful in analytical chemistry, biology, and pharmaceutical fields
  • Highly permeable to water and low molecular weight compounds
  • Contains glycerin, water and 1% sulfur
  • Must be stored in airtight container or rehumidified before use
  • Pore diameter: 4.8nm

This product is for educational use - no C of A is available.

Specifications

Specifications

Dialysis Tubing
20.4 mm
10 ft.
Glycerin, water and 1% sulfur
Useful in analytical chemistry, biology, and pharmaceutical fields
Seamless Cellulose
0.8 in.
3 m
Each
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers