missing translation for 'onlineSavingsMsg'
Learn More

Sebacoyl chloride, 92%, technical

Product Code. 10422261 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
50 g
250 g
500 g
Packaging:
Glass bottle
Unit Size:
250g
500g
50g
This item is not returnable. View return policy

Product Code. 10422261

Brand: Thermo Scientific Acros 294902500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sebacoyl chloride, 92%, C10H16Cl2O2, CAS Number-111-19-3, decanedioic acid chloride, sebacoyl chloride, cp, octane-1,8-dicarbonyl chloride, sebacyl chloride, sebacoyl dichloride, sebacic acid dichloride, sebacoyl chloride, sebacoylchloride, acmc-1byrk, decanedioyl chloride, 500g, 90% min. (GC) | CAS: 111-19-3 | C10H16Cl2O2 | 239.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-19-3
Molecular Formula C10H16Cl2O2
Molecular Weight (g/mol) 239.14
MDL Number MFCD00000770
InChI Key WMPOZLHMGVKUEJ-UHFFFAOYSA-N
Synonym sebacoyl chloride, sebacyl chloride, sebacoyl dichloride, sebacic acid dichloride, decanedioyl chloride, sebacoylchloride, octane-1,8-dicarbonyl chloride, sebacoyl chloride, cp, acmc-1byrk, decanedioic acid chloride
PubChem CID 66072
IUPAC Name decanedioyl dichloride
SMILES C(CCCCC(=O)Cl)CCCC(=O)Cl

Specifications

Melting Point -2.5°C
Color White to Beige
Density 1.1210g/mL
Boiling Point 220.0°C (75.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 90% min. (GC)
Packaging Glass bottle
Linear Formula ClCO(CH2)8COCl
Refractive Index 1.4670 to 1.4700
Quantity 250 g
Beilstein 02, 719
Specific Gravity 1.121
Solubility Information Solubility in water: may decompose. Other solubilities: soluble in hydrocarbons and ethers
Formula Weight 239.14
Percent Purity 92%
Grade Technical
Physical Form Liquid
Chemical Name or Material Sebacoyl chloride
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Sebacoyl chloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
Hazard Statement
  • H290-May be corrosive to metals.
  • H302-Harmful if swallowed.
  • H310-Fatal in contact with skin.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Decanedioyl dichloride, Hydrochloric acid, Decanedioic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.