missing translation for 'onlineSavingsMsg'
Learn More

Sebacoyl chloride, 95%, Thermo Scientific Chemicals

Product Code. 11412337
25 g
Click to view available options
Quantity:
25 g
100 g
500 g

Sebacoyl chloride is used in the synthesis of nylon-6,10 polymer by reacting with hexamethylenediamine. It is also used to prepare decanedioic acid bis-(3-dimethylamino-propyl) ester in the presence of triethylamine as catalyst. It is used as an intermediate in chemical synthesis. It is involved in the production of polyamide microcapsules by polycondensation of p-phenylenediamine with sebacoyl chloride using diethylenetriamine (DETA) cross-linker and oleic acid as a plasticizer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sebacoyl chloride is used in the synthesis of nylon-6,10 polymer by reacting with hexamethylenediamine. It is also used to prepare decanedioic acid bis-(3-dimethylamino-propyl) ester in the presence of triethylamine as catalyst. It is used as an intermediate in chemical synthesis. It is involved in the production of polyamide microcapsules by polycondensation of p-phenylenediamine with sebacoyl chloride using diethylenetriamine (DETA) cross-linker and oleic acid as a plasticizer.

Solubility
Miscible with hydrocarbons and ethers.

Notes
Store in a cool place. Moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with water, acids, bases, amines, alcohols and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-19-3
Molecular Formula C10H16Cl2O2
Molecular Weight (g/mol) 239.136
MDL Number MFCD00000770
InChI Key WMPOZLHMGVKUEJ-UHFFFAOYSA-N
Synonym sebacoyl chloride, sebacyl chloride, sebacoyl dichloride, sebacic acid dichloride, decanedioyl chloride, sebacoylchloride, octane-1,8-dicarbonyl chloride, sebacoyl chloride, cp, acmc-1byrk, decanedioic acid chloride
PubChem CID 66072
IUPAC Name decanedioyl dichloride
SMILES C(CCCCC(=O)Cl)CCCC(=O)Cl

Specifications

Melting Point -5°C
Density 1.121
Boiling Point 161°C (14 mmHg)
Flash Point 165°C (329°F)
Odor Acrid
Refractive Index 1.468
Quantity 25 g
UN Number UN2922
Beilstein 1365665
Sensitivity Moisture sensitive
Solubility Information Miscible with hydrocarbons and ethers.
Formula Weight 239.14
Percent Purity 97%
Chemical Name or Material Sebacoyl chloride
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Sebacoyl chloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H302-Harmful if swallowed.
  • H310-Fatal in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Decanedioyl dichloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sebacoyl chloride, 95%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.