missing translation for 'onlineSavingsMsg'
Learn More

Sebacoyl Chloride, Extra Pure, 5% Solution In Trichloroethylene, SLR, Fisher Chemical

Product Code. 10529580 Shop All Fisher Chemical Products
Click to view available options
Quantity:
100 mL
Packaging:
Plastic Coated Glass bottle
Unit Size:
100mL
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Product Code. 10529580

Brand: Fisher Chemical S/0423/PB05

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Specifications

CAS 79-01-6
Melting Point -2.5°C
Color Colorless
Density 1.121g/mL at 25°C
Boiling Point 220°C
Flash Point >230°F
Packaging Plastic Coated Glass bottle
Molecular Formula C2HCl3
Refractive Index n20/D 1.468
MDL Number 770
Quantity 100 mL
UN Number 1710
Sensitivity Moisture sensitive
Synonym trichloroethylene, trichloroethene, ethene, trichloro, trilene, trichlorethylene, ethinyl trichloride, ethylene trichloride, acetylene trichloride, chlorilen, narcogen
Solubility Information Reacts violently with water liberating toxic gas. Incompatible with water, oxidizing agents, amines, alcohols.
InChI Key XSTXAVWGXDQKEL-UHFFFAOYSA-N
SMILES C(=C(Cl)Cl)Cl
IUPAC Name 1,1,2-trichloroethene
Molecular Weight (g/mol) 131.38
PubChem CID 6575
ChEBI CHEBI:16602
Formula Weight 239.14
Grade Extra Pure
Concentration or Composition (by Analyte or Components) 5% Solution in Trichloroethylene
Physical Form Liquid
Chemical Name or Material Sebacoyl Chloride, 5% Solution in Trichloroethylene
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Sebacoyl chloride solution 5% in trichloroethylene
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
  • Germ cell mutagenicity Category 2
  • Carcinogenicity Category 1B
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H312-Harmful in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H336-May cause drowsiness or dizziness.
  • H341-Suspected of causing genetic defects.
  • H350-May cause cancer.
  • H412-Harmful to aquatic life with long lasting effects.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Trichloroethylene, Decanedioyl dichloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.