missing translation for 'onlineSavingsMsg'
Learn More

sec-Butanol, 99+%, for analysis, Thermo Scientific Chemicals

Product Code. 10590594
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-92-2
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.12
MDL Number MFCD00004569
InChI Key BTANRVKWQNVYAZ-UHFFFAOYNA-N
Synonym 2-butanol, sec-butanol, sec-butyl alcohol, 2-hydroxybutane, 2-butyl alcohol, s-butyl alcohol, butylene hydrate, s-butanol, 1-methyl-1-propanol, 1-methylpropyl alcohol
PubChem CID 6568
ChEBI CHEBI:35687
IUPAC Name butan-2-ol
SMILES CCC(C)O

Specifications

Melting Point -115.0°C
Density 0.8100g/mL
Boiling Point 99.0°C
Quantity 2.5 L
Flash Point 24°C
Packaging Glass Bottle
Physical Form Liquid
Assay Percent Range 99+%
Specific Gravity 0.81
Linear Formula C2H5CH(OH)CH3
Viscosity 4.2 mPa.s (20°C)
Beilstein 01, 371
Water 0.1% max.
Merck Index 15, 1543
Solubility Information Solubility in water: 125g/L (20°C). Other solubilities: soluble in alcohol, ester, ether and benzene
Formula Weight 74.12
Percent Purity ≥99%
Grade Analytical
Chemical Name or Material sec-Butanol, For analysis
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.