missing translation for 'onlineSavingsMsg'
Learn More

sec-Butanol, 99%, Extra Pure, Thermo Scientific Chemicals

Product Code. 10512744
Click to view available options
Quantity:
1 L
2.5 L
10 L
Packaging:
Glass Bottle
Plastic Drum

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-92-2
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.12
MDL Number MFCD00004569
InChI Key BTANRVKWQNVYAZ-UHFFFAOYNA-N
Synonym 2-butanol, sec-butanol, sec-butyl alcohol, 2-hydroxybutane, 2-butyl alcohol, s-butyl alcohol, butylene hydrate, s-butanol, 1-methyl-1-propanol, 1-methylpropyl alcohol
PubChem CID 6568
ChEBI CHEBI:35687
IUPAC Name butan-2-ol
SMILES CCC(C)O

Specifications

Melting Point -115.0°C
Density 0.8100g/mL
Boiling Point 99.0°C
Quantity 10 L
Flash Point 24°C
Infrared Spectrum Conforms to Ref Report
Packaging Plastic Drum
Physical Form Liquid
Refractive Index 1.3960 to 1.3980 (20°C, 589nm)
Assay Percent Range 99%
Residue after Evaporation 0.005% max.
Specific Gravity 0.81
Linear Formula C2H5CH(OH)CH3
Viscosity 4.2 mPa.s (20°C)
Beilstein 01, 371
Water 0.5% max.
Merck Index 15, 1543
Solubility Information Solubility in water: 125g/L (20°C). Other solubilities: soluble in alcohol, ester, ether and benzene
Formula Weight 74.12
Percent Purity 99%
Grade Extra Pure
Chemical Name or Material sec-Butanol
Mehr anzeigen Weniger anzeigen

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'