missing translation for 'onlineSavingsMsg'
Learn More
sec-Butyllithium, 1.3M sol. in cyclohexane/hexane (92/8), AcroSeal™, Thermo Scientific Chemicals Product Code.: 10568513

sec-Butyllithium, 1.3M sol. in cyclohexane/hexane (92/8), AcroSeal™, Thermo Scientific Chemicals

Product Code. 10568513
800 mL, AcroSeal™ Glass Bottle
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 
Sea freight only: For EU stock allow up to 15 days delivery.
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

Product Code. 10568513

Brand: Thermo Scientific Acros 187548000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 
Sea freight only: For EU stock allow up to 15 days delivery.
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 598-30-1, 110-82-7, 110-54-3
Molecular Formula C4H9Li
Molecular Weight (g/mol) 64.06
MDL Number MFCD00009323
InChI Key VATDYQWILMGLEW-UHFFFAOYNA-N
Synonym sec-butyllithium, s-buli, s-butyllithium, s-butyl lithium, unii-5yv3gii1tb, lithium, 1-methylpropyl, 5yv3gii1tb, secbutyllithium, b-butyllithium, sec-butyllitium
PubChem CID 102446
IUPAC Name (butan-2-yl)lithium
SMILES [Li]C(C)CC

Specifications

Color Colorless
Linear Formula C2H5CH(CH3)Li
Fieser 09,87; 10,75; 12,97; 16,56
Density 0.7500g/mL
Solubility Information Solubility in water: reacts (forming lioh and butane).
Formula Weight 64.06
Percent Purity 10.5 to 12.5% (as BuLi) (Active base)
Packaging AcroSeal™ Glass Bottle
Specific Gravity 0.75
Quantity 800 mL
Flash Point −17°C
Physical Form Liquid
Chemical Name or Material sec-Butyllithium, 1.3M Solution in Cyclohexane/hexane (92/8)
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0467157 Certificate TypeCertificate of Analysis Date31/10/2024 Catalog Number187540090,187541000,187548000
Lot NumberA0455961 Certificate TypeCertificate of Analysis Date29/04/2024 Catalog Number187540090,187541000,187548000
Lot NumberA0448866 Certificate TypeCertificate of Analysis Date11/03/2024 Catalog Number187540090,187541000,187548000
Lot NumberA0452515 Certificate TypeCertificate of Analysis Date31/01/2024 Catalog Number187541000,187548000
Lot NumberA0394983 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number187540090,187541000,187548000
compliance-icons compliance-icons compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • Aspiration toxicity Category 1
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Pyrophoric liquid Category 1
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
  • Substance which\, in contact with water\, emits flammable gases Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H250-Catches fire spontaneously if exposed to air.
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • H304-May be fatal if swallowed and enters airways.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H336-May cause drowsiness or dizziness.
  • H361f-Suspected of damaging fertility.
  • H373-May cause damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P222-Do not allow contact with air.
  • P231+P232-Handle under inert gas. Protect from moisture.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
sec-Butyllithium, 1.3M sol. in cyclohexane/hexane (92/8), AcroSeal™, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.