Learn More
SEC24D Antibody (2D4), Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals H00009871-M03
Product Code. 18318079
  • £469.00 / 0.01mg
Explore more special offers
Description
Description
SEC24D Monoclonal antibody specifically detects SEC24D in Human samples. It is validated for Western Blot, ELISA
Specifications
Specifications
SEC24D
Monoclonal
Unconjugated
In 1x PBS, pH 7.4
FLJ43974, KIAA0755member D, SEC24 family, member D (S. cerevisiae), SEC24 related gene family, member D
SEC24D (NP_055637, 935 a.a. ~ 1032 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. SPPELIQGIFNVPSFAHINTDMTLLPEVGNPYSQQLRMIMGIIQQKRPYSMKLTIVKQREQPEMVFRQFLVEDKGLYGGSSYVDFLCCVHKEICQLLN
0.1 mg
Primary
Human
Purified
Western Blot, ELISA
2D4
Western Blot, ELISA
NP_055637
Mouse
IgG purified
RUO
9871
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
IgG1 κ