missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Secnidazole, Thermo Scientific™
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Specifications
Specifications
| Chemical Name or Material | Secnidazole |
| Type | Secnidazole |
| Physical Form | Powder |
| CAS | 3366-95-8 |
| Melting Point | 70°C to 76°C |
| Infrared Spectrum | Conforms |
| Packaging | Glass Bottle |
| Water | ≤6 % |
| Color | White to Yellow |
| Quantity | 5 g |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction