missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Second Strand cDNA Synthesis Kit

Invitrogen™ Second Strand cDNA Synthesis Kit is design to produce double-stranded cDNA using first strand reaction without the need for intermediate organic extraction or ethanol precipitation steps. This convenient single-tube format speeds up the synthesis procedure and maximizes cDNA recovery. The kit contains premixed components to reduce the number of pipetting steps necessary to complete the procedure.

Brand:  Invitrogen™ A48570

771.40 GBP valid until 2024-12-31
Use promo code "22010" to get your promotional price.


 View more versions of this product

Product Code. 17195802

  • £812.00 / 50 reactions

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

InvitrogenTM Second Strand cDNA Synthesis Kit is design to produce double-stranded cDNA using first strand reaction without the need for intermediate organic extraction or ethanol precipitation steps. This convenient single-tube format speeds up the synthesis procedure and maximizes cDNA recovery. The kit contains premixed components to reduce the number of pipetting steps necessary to complete the procedure.

Features of the Second Strand cDNA Synthesis Kit include:

  • Efficient synthesis of full-length double-stranded cDNA
  • Fast—procedure completed in 1 hour
  • Convenient—pre-mixed components
  • Complete—includes residual RNA removal reagents

Applications

  • Full-length double-stranded blunt-end cDNA synthesis for cloning
  • Double-stranded cDNA library construction
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Contents:
• Second Strand Enzyme Mix, 1 mL
• 5X Second Strand Reaction Mix, 250 μL
• 0.5 M EDTA, pH 8.0, 1 mL
• RNase I, 10 U/μL, 500 μL
• Water, nuclease-free, 3 x 1.25 mL

Storage: -15 to -25°C

cDNA Synthesis Kit
Cloning
50 Reactions
50 reactions
50 reaction
Double Stranded cDNA
Separate Components
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.