Learn More
3M™ Secure Click™ Half Facepiece Reusable Respirator
3M™ Secure Click™ Half Facepiece Reusable Respirator is designed with smart and intuitive features, which makes it simple and reliable.
Brand: 3M™ 7100172003
This item is not returnable.
View return policy
Description
- Greater breathability and comfort provided by the world's first quad flow cartridge system.
- Push button seal check helps bolster confidence that the respirator is properly sealed to the face.
- Speaking diaphragm is designed to help provide easier communication while you work.
- Confidence that filters and cartridges are installed properly provided by all new Secure Click™ connection.
- Low profile half mask design offers a wide field of view and compatibility with certain welding and grinding shields.
- Over-moulded design for easy assembly/disassembly.
- Advanced silicone material for increased comfort and greater durability.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
AS/NZS 1716:2012 | |
Dark Gray | |
Half Facepiece Respirator | |
Without Speaking Diaphragm | |
10/Cs. |
Secure Click™ | |
Silicone, Nylon | |
Medium | |
HF-802 |
Product Content Correction
Product Title
3M™ Secure Click™ Half Facepiece Reusable Respirator > Half Mask Reusable Respirator; Size: Medium; Includes: Without Speaking Diaphragm
Spot an opportunity for improvement?Share a Content Correction