Learn More
3M™ Secure Click™ Hard Case P3 R Particulate Filter, D9035
3M™ Secure Click™ Hard Case Particulate Filter for use with 3M™ Secure Click™ Respirators to provide protection against exposure to particles, dusts, and mists in environments containing certain particles.
Brand: 3M™ 7100206891
97.70 GBP valid until 2024-12-31
Use promo code "22912" to get your promotional price.
Additional Details : Weight : 3.48000kg
Description
- For use with 3M™ Secure Click™ Respirators
- Filter encapsulated in lightweight plastic case
- Hard case recommended for harsh environments with a need for easy decontamination and protection
- Hard case helps protect against sparks and high heat
- Filter case designed for certain spray applications
- Secure Click™ connection allows filters to be easily and securely inserted into place
- Solid top and channelled airflow help protect filter
- Swept-back design provides wide field of view
- CE Approved
Specifications
P | |
Filter | |
3M™ Secure Click™ Half Facepiece Reusable Respirators HF-800 Series | |
40/Pk. |