Learn More
3M™ Secure Click™ Reusable Full Face Respirator
The Secure Click™ Reusable Full Face Mask FF-800 Series helps protect against particulates and a wide variety of gases and vapors when used with approved Secure Click™ filters.
Brand: 3M™ 7100262626
221.00 GBP valid until 2024-12-31
Use promo code "22912" to get your promotional price.
Additional Details : Weight : 0.64047kg
Description
- The quick, easy, one-touch positive pressure push button seal check helps provide improved confidence in respirator fit
- Two dual-flow gas and vapor filters on each respirator combine to create the world's first quad flow gas and vapor filter system with four airflow paths providing easy breathing and comfort
- Large lens for excellent field of vision, and treated with Scotchgard™ Protector for improved paint and stain resistance
- Six-strap harness helps provide a secure comfortable fit
- Speaking diaphragm is designed to help provide easy communication
- Recommended applications include assembly, chemical clean-up, chemical handling, chipping, chiseling, cleaning, furnace operations, grinding, machining, masonry, painting, pouring/casting, sanding and sawing.
- Available in three sizes
Specifications
Black | |
Secure Click™ Filters | |
Large |