Learn More
3M™ SecureFit™ 400X Safety Glasses, Scotchgard™ Anti-Fog / Anti-Scratch Coating (K&N) Product Code.: 16476380
In Stock
to see stock.
Add to basket
3M™ SecureFit™ 400X Safety Glasses, Scotchgard™ Anti-Fog / Anti-Scratch Coating (K&N)
Product Code. 16476380
Yellow, Black, Amber
Click to view available options
Frame Color:
Black
Blue, Gray
Green, Black
Green, Clear
Yellow, Black
Lens Tint:
Amber
Clear
Gray
Light Gray, Indoor/Outdoor
Orange, Mirror
Vermillion
Product Code. 16476380
Brand: 3M™ 7100185291
In Stock
to see stock.
Edge
Add to basket
Add to basket
3M™ SecureFit™ 400X Safety Glasses have 3M™ Pressure Diffusion Temple Technology, flexible temple arms and combined soft nose bridge/brow guard. Lightweight anti-scratch & superior anti-fog, rimless polycarbonate lens for indoor/outdoor use.
  • Safety glasses with 3M™ Pressure Diffusion Temple Technology for comfort
  • Wraparound design to improve the security of fit and prevent slipping
  • Padded temple touchpoints for increased comfort over the ears
  • Soft nose bridge for a more comfortable fit
  • Contoured brow guard provides additional coverage from flying particles
  • Superior anti-fog and anti-scratch properties meets the K&N marking requirements of EN166
  • Frame marking: SF400 Series, 3M EN166 FT
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance CE Approved
Lens Coating Anti-fog
Lens Tint Amber
Product Type Protective Eyewear
Frame Color Yellow, Black
Temple Style Pressure Diffusion Temple Technology
Lens Material Polycarbonate
Frame Material Plastic
UV Protection UV400
Product Content Correction
Product Title
3M™ SecureFit™ 400X Safety Glasses, Scotchgard™ Anti-Fog / Anti-Scratch Coating (K&N) >