Learn More
3M™ SecureFit™ SF600 Series Safety Glasses
Product Code. 15727727
Click to view available options
Lens Coating:
Anti-fog, Scratch-resistant
Scratch-resistant
Lens Tint:
Amber
Clear
Gray
I/O Mirror
Welding shade 1.7
Welding shade 3.0
Welding shade 5.0
Unit Size:
Each
Lens Type:
Polarized
Product Code. 15727727
Brand: 3M™ 7100112718
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Comprised of a semi-rimless lens and fixed-length temple arm. 3M™ SecureFit™ SF600 Series Safety Glasses include an integrated side shield for added protection.
  • Optical class 1 lens suitable for prolonged use
  • Design provides coverage and a good field of vision based on two lenses in a semi-rimless design
  • Offers protection against Ultraviolet (UV) radiation in accordance with European standards
  • Weight: 27g
  • 3M™ Pressure Diffusion Temple Technology for secure and comfortable fit
  • Soft nose pads for comfortable fit
  • Superior Scotchgard™ anti-fog coating meeting K and N on the standard clear, gray, and amber lenses
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Scratch-resistant
Lens Tint I/O Mirror
Frame Color Gray
Lens Material Polycarbonate
Frame Material Polycarbonate
UV Protection Yes