missing translation for 'onlineSavingsMsg'
Learn More

3M™ SecureFit™ X5000 Safety Helmet, Vented, 1000 V, CE

3M™ SecureFit™ Safety Helmet has six different settings to adjust position and a 4-point chin strap selector system. Vented and brimless safety hat with 3M™ UVicator™ Sensor that indicates damaging levels of UV light.

Brand:  3M™ 7100175536

285.28 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 17315333

  • £505.00 / Pack of 4
Estimated Shipment: 30-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Secure, comfortable head protection for working at heights and other hazardous industrial jobs
  • Suspension system incorporates exclusive patented 3M™ Pressure Diffusion Technology
  • 4-point chin strap selector system unique to 3M allows wearer to switch between EN 12492 (helmets for mountaineers) and EN 397 (industrial safety helmets) standards
  • 6-point ratchet suspension allows wearer to customise height and fit with multiple levels of vertical adjustment
  • Inspired by climbing helmets with a brimless design to give you a clear upward view
  • Accessory slots and clips are compatible with a variety of 3M face shields, earmuffs and other accessories
  • 3M™ Uvicator™ sensor disc tells you when your hard hat has been over exposed to UV-radiation and it's time to replace it
  • Certified to either EN12492 or EN397 (depending on chin strap configuration) and EN50365 1000 Vac standard
  • Approved for lateral deformation (LD)
  • UV-stabilized ABS
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Black
ABS
Pressure Diffusion 6 Point Ratchet
Construction, Forestry, Oil, and Gas
Safety Helmet
Ventilated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers