missing translation for 'onlineSavingsMsg'
Learn More

3M™ SecureFit™ X5000 Safety Helmet, Vented, 1000 V, CE

3M™ SecureFit™ Safety Helmet has six different settings to adjust position and a 4-point chin strap selector system. Vented and brimless safety hat with 3M™ UVicator™ Sensor that indicates damaging levels of UV light.

Brand:  3M™ 7100175512

285.28 GBP valid until 2024-12-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 17375323

  • £505.00 / Pack of 4
Estimated Shipment: 05-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Secure, comfortable head protection for working at heights and other hazardous industrial jobs
  • Suspension system incorporates exclusive patented 3M™ Pressure Diffusion Technology
  • 4-point chin strap selector system unique to 3M allows wearer to switch between EN 12492 (helmets for mountaineers) and EN 397 (industrial safety helmets) standards
  • 6-point ratchet suspension allows wearer to customise height and fit with multiple levels of vertical adjustment
  • Inspired by climbing helmets with a brimless design to give you a clear upward view
  • Accessory slots and clips are compatible with a variety of 3M face shields, earmuffs and other accessories
  • 3M™ Uvicator™ sensor disc tells you when your hard hat has been over exposed to UV-radiation and it's time to replace it
  • Certified to either EN12492 or EN397 (depending on chin strap configuration) and EN50365 1000 Vac standard
  • Approved for lateral deformation (LD)
  • UV-stabilized ABS
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Blue
ABS
Pressure Diffusion 6 Point Ratchet
Construction, Forestry, Oil, and Gas
Safety Helmet
Ventilated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.