Learn More
3M™ SecureFit™ X5000 Safety Helmet, Vented, 1000 V, CE
3M™ SecureFit™ Safety Helmet has six different settings to adjust position and a 4-point chin strap selector system. Vented and brimless safety hat with 3M™ UVicator™ Sensor that indicates damaging levels of UV light.
Brand: 3M™ 7100175533
274.00 GBP valid until 2024-12-31
Use promo code "22912" to get your promotional price.
Additional Details : Weight : 2.02320kg
Description
- Secure, comfortable head protection for working at heights and other hazardous industrial jobs
- Suspension system incorporates exclusive patented 3M™ Pressure Diffusion Technology
- 4-point chin strap selector system unique to 3M allows wearer to switch between EN 12492 (helmets for mountaineers) and EN 397 (industrial safety helmets) standards
- 6-point ratchet suspension allows wearer to customise height and fit with multiple levels of vertical adjustment
- Inspired by climbing helmets with a brimless design to give you a clear upward view
- Accessory slots and clips are compatible with a variety of 3M face shields, earmuffs and other accessories
- 3M™ Uvicator™ sensor disc tells you when your hard hat has been over exposed to UV-radiation and it's time to replace it
- Certified to either EN12492 or EN397 (depending on chin strap configuration) and EN50365 1000 Vac standard
- Approved for lateral deformation (LD)
- UV-stabilized ABS
Specifications
Green | |
ABS | |
Pressure Diffusion 6 Point Ratchet |
Construction, Forestry, Oil, and Gas | |
Safety Helmet | |
Ventilated |