missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lenz Laborglasintrumente Sedimentation Cone Acc. to Imhoff with Stopcock
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand: Lenz Laborglasintrumente 05670005
This item is not returnable.
View return policy
Description
- Sedimentation cone acc. to DIN 12672
- Scale: up to 100 mL
- Ring mark: at 1000 mL
- Subdivision: 0.1 ml (0-2 mL), 0.5 mL (2-10 mL), 1 ml (10-40 mL), 2 mL (20-100 mL)
- Material: borosilicate glass 3.3
Specifications
Borosilicate Glass 3.3 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction