Learn More
3M™ Petrifilm™ Select E. Coliplate
Specific for E. coli, only E. coli colonies are visible which numerate within 24 hours
Brand:  3M™ Petrifilm™ 7100126813
Encompass
 View more versions of this product
Product Code. 17223784
  • £836.73 / Pack of 500
Estimated Shipment: 13-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Specific test for E. coli, only E. coli colonies are visible
  • E. coli enumerates in just 24 hours
  • Contains a ß-glucuronidase indicator
Specifications
Specifications
E. coli
Microbiology Media
Food and Beverage, Environmental Test
500 Pack