missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Selenium, 99+%, powder, Thermo Scientific Chemicals
44.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Use promo code "22914" to get your promotional price.
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Boiling Point | 685°C |
Melting Point | 217°C |
Color | Cream-White to White |
Physical Form | Powder |
Quantity | 100 g |
Assay Percent Range | 99% min |
Linear Formula | Se |
Fieser | 01,990; 04,422; 05,575; 06,507; 08,439; 10,345; 11,465; 14,281 |
Merck Index | 15, 8568 |
Solubility Information | Solubility in water: insoluble. Other solubilities: soluble in concentrated acid,slightly soluble in diehtyl ether,almost insoluble in carbon disulfide,insoluble in ethanol |
Show More |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS06_Toxic.png)
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS08_Dangerous_Health_Hazard.png)
Product Identifier
- Selenium
- Danger
- Acute toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 4
- Specific target organ toxicity after repeated exposure Category 2
- H373-May cause damage to organs.
- H331-Toxic if inhaled.
- H413-May cause long lasting harmful effects to aquatic life.
- H301-Toxic if swallowed.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'