missing translation for 'onlineSavingsMsg'
Learn More

Selenium AAS Standard, Fluka™

Certified Reference Material, 1000 mg/L Se in HNO3 2%

Brand:  Honeywell Fluka E4067-100ML

Additional Details : CAS Number : 7697-37-2 Weight : 0.10000kg

 View more versions of this product

Product Code. 16666232

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

Honeywell Fluka™ analytical standards offers a growing range of high quality inorganic and organic reference materials for the European market produced according to ISO 9001. Our products are suitable for a wide range of industries and applications and come complete with a detailed Certificate of Analysis (CoA) in every delivery, and are eligible for free expert technical support ensuring you have the information you need to quickly start your analysis and pass an audit.

Specifications

Specifications

Selenium AAS Standard
7783-00-8
AAS
70°C (Decomposition)
Plastic Bottle
100 mL
<2
684.9°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers