missing translation for 'onlineSavingsMsg'
Learn More

Selenium(IV) oxide, 99.8%, (trace metal basis), Thermo Scientific Chemicals

£26.03 - £985.91

Chemical Identifiers

CAS 7446-08-4
Molecular Formula O2Se
Molecular Weight (g/mol) 110.97
MDL Number MFCD00003562
InChI Key JPJALAQPGMAKDF-UHFFFAOYSA-N
Synonym selenium oxide, selenium iv oxide, oxoselane oxide, selenious anhydride, selenium oxide seo2, seleniumdioxide, dimer, selenous acid anhydride, selenium iv dioxide 1:2, selenium oxide se2o4
PubChem CID 24007
SMILES O=[Se]=O
View More Specs

Products 4
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10266540
View Documents Promotion Details
Thermo Scientific Acros
200000100
10 g Glass bottle
£26.03
10g
Please to purchase this item. Need a web account? Register with us today!
10665082
View Documents
Thermo Scientific Acros
200001000
100 g Glass bottle
£72.46
100g
Please to purchase this item. Need a web account? Register with us today!
10480281
View Documents
Thermo Scientific Acros
200005000
500 g Glass bottle
£268.34
500g
Please to purchase this item. Need a web account? Register with us today!
11964121
View Documents
Thermo Scientific Acros
200000025
2.5 kg Glass bottle
£985.91
2.50kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7446-08-4
110.97
JPJALAQPGMAKDF-UHFFFAOYSA-N
24007
O2Se
MFCD00003562
selenium oxide, selenium iv oxide, oxoselane oxide, selenious anhydride, selenium oxide seo2, seleniumdioxide, dimer, selenous acid anhydride, selenium iv dioxide 1:2, selenium oxide se2o4
O=[Se]=O

Specifications

315.0°C
White to Beige
70 to 71.5% Se
SeO2
01,992; 02,360; 03,245; 04,422; 05,575; 06,509; 08,439; 09,409; 10,345; 11,96; 13,272; 14,177; 15,89; 17,312
selenium oxide, selenium iv oxide, oxoselane oxide, selenious anhydride, selenium oxide seo2, seleniumdioxide, dimer, selenous acid anhydride, selenium iv dioxide 1:2, selenium oxide se2o4
JPJALAQPGMAKDF-UHFFFAOYSA-N
selenium dioxide
24007
99.8%
Selenium(IV) oxide
7446-08-4
Powder
O2Se
MFCD00003562
15,8571
Solubility in water: 38.4 g/100mL (14°C). Other solubilities: 1.11 parts/100 parts acetic acid (13.9°C), 10.16 parts/100 parts methanol (11.8°C), 6.67 parts/100 parts 93% ethanol (14°C), 4.35 parts/100 parts acetone (15.3°C)
O=[Se]=O
110.97
110.96
Trace Metal Basis
Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 231-194-7

Product Identifier
  • Selenium(IV) oxide

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Specific target organ toxicity after repeated exposure Category 2

Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H373-May cause damage to organs.
  • H331-Toxic if inhaled.
  • H301-Toxic if swallowed.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only