Learn More
Selenium(IV) oxide, 99.999%, (trace metal basis), Thermo Scientific Chemicals
£97.99 - £385.92
Chemical Identifiers
CAS | 7446-08-4 |
---|---|
Molecular Formula | O2Se |
Molecular Weight (g/mol) | 110.97 |
MDL Number | MFCD00003562 |
InChI Key | JPJALAQPGMAKDF-UHFFFAOYSA-N |
Synonym | selenium oxide, selenium iv oxide, oxoselane oxide, selenious anhydride, selenium oxide seo2, seleniumdioxide, dimer, selenous acid anhydride, selenium iv dioxide 1:2, selenium oxide se2o4 |
PubChem CID | 24007 |
IUPAC Name | selenium dioxide |
SMILES | O=[Se]=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10358620
|
Thermo Scientific Acros
193980100 |
10 g | Glass bottle |
£97.99
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10286820
|
Thermo Scientific Acros
193980500 |
50 g | Glass bottle |
£385.92
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysisChemical Identifiers
7446-08-4 | |
110.97 | |
JPJALAQPGMAKDF-UHFFFAOYSA-N | |
24007 | |
O=[Se]=O |
O2Se | |
MFCD00003562 | |
selenium oxide, selenium iv oxide, oxoselane oxide, selenious anhydride, selenium oxide seo2, seleniumdioxide, dimer, selenous acid anhydride, selenium iv dioxide 1:2, selenium oxide se2o4 | |
selenium dioxide |
Specifications
315°C | |
White | |
99.999% | |
SeO2 | |
01,992; 02,360; 03,245; 04,422; 05,575; 06,509; 08,439; 09,409; 10,345; 11,96; 13,272; 14,177; 15,89; 17,312 | |
selenium oxide, selenium iv oxide, oxoselane oxide, selenious anhydride, selenium oxide seo2, seleniumdioxide, dimer, selenous acid anhydride, selenium iv dioxide 1:2, selenium oxide se2o4 | |
JPJALAQPGMAKDF-UHFFFAOYSA-N | |
selenium dioxide | |
24007 | |
100.00% | |
Selenium(IV) oxide, 99.999% |
7446-08-4 | |
Fine Crystalline Powder | |
O2Se | |
MFCD00003562 | |
15, 8571 | |
Solubility in water: 38.4g/100mL (14°C). Other solubilities: 1.11 parts/100 parts acetic acid (13.9°C),10.16 parts/100 parts methanol (11.8°C),6.67 parts/100 parts 93% ethanol (14°C),4.35 parts/100 parts acetone (15.3°C) | |
O=[Se]=O | |
110.97 | |
110.96 | |
Trace Metal Basis |
Safety and Handling
GHS H Statement:
Toxic if swallowed.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Very toxic to aquatic life with long lasting effects.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 231-194-7
missing translation for 'rtecsNumber' : VS8575000
missing translation for 'tsca' : TSCA
- Selenium(IV) oxide
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 2
- Acute toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H301-Toxic if swallowed.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H330-Fatal if inhaled.
- H373-May cause damage to organs.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.