Learn More
Selenium, powder, 99+%, Thermo Scientific Chemicals
Brand: Thermo Scientific Alfa Aesar A12592.30
Code : NEW
Additional Details : CAS Number : 7782-49-2 Weight : 0.25000kg Transport : UN number : 3288 Chem class : 6.1 Pack group : III
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
690°C | |
224°C to 225°C | |
250 g | |
≥99% |
Safety and Handling
- Selenium powder
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 4
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H301-Toxic if swallowed.
- H331-Toxic if inhaled.
- H373-May cause damage to organs.
- H413-May cause long lasting harmful effects to aquatic life.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P311-Call a POISON CENTER/doctor/ .
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- P405-Store locked up.
Supplemental information
- MIXTURE LIST-Contains : Selenium